Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TLE3 Monoclonal Antibody (CL3573)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524657
Description
Immunogen sequence: GSQAHLTVKD EKNHHELDHR ERESSANNSV SPSESLRASE KHR Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.
Specifications
| TLE3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q04726 | |
| TLE3 | |
| Recombinant protein corresponding to Human TLE3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL3573 | |
| Unconjugated | |
| TLE3 | |
| 2610103N05Rik; enhancer of split groucho 3; enhancer of split groucho-like protein 3; ESG; ESG-1; ESG-2; ESG3; ESG-3; ESG-4; Esp3; GRG3; Grg-3; Grg3a; Grg3b; HsT18976; KIAA1547; mKIAA1547; rTLE3; TLE family member 3, transcriptional corepressor; Tle1; Tle2; Tle3; tle4; transducin like enhancer of split 3; transducin-like enhancer of split 3; transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 3 homolog of Drosophila; transducin-like enhancer of split 3 short isoform; transducin-like enhancer of split 3, E(spl) homolog; transducin-like enhancer of split 3, homolog of Drosophila E(sp1); transducin-like enhancer of split 3, homolog of Drosophila E(spl); transducin-like enhancer protein 3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7090 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction