Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540528 Supplier Invitrogen™ Supplier No. PA540528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GSSSGLLALG ALGSQAHLTV KDEKNHHELD HRERESSANN SVSPSESLRA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 93%; Zebrafish: 79%.

The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLE3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TLE3
Gene Accession No. Q04726
Gene Alias 2610103N05Rik; enhancer of split groucho 3; enhancer of split groucho-like protein 3; ESG; ESG-1; ESG-2; ESG3; ESG-3; ESG-4; Esp3; GRG3; Grg-3; Grg3a; Grg3b; HsT18976; KIAA1547; mKIAA1547; rTLE3; TLE family member 3, transcriptional corepressor; Tle1; Tle2; Tle3; tle4; transducin like enhancer of split 3; transducin-like enhancer of split 3; transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 3 homolog of Drosophila; transducin-like enhancer of split 3 short isoform; transducin-like enhancer of split 3, E(spl) homolog; transducin-like enhancer of split 3, homolog of Drosophila E(sp1); transducin-like enhancer of split 3, homolog of Drosophila E(spl); transducin-like enhancer protein 3
Gene Symbols TLE3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TLE3 (aa 161-210).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7090
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.