Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLE4 Monoclonal Antibody (384CT24.1.3.4)
GREENER_CHOICE

Catalog No. PIMA537554
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537554 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537554 Supplier Invitrogen™ Supplier No. MA537554
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLE4 Monoclonal Antibody for Western Blot

Transcriptional corepressor that binds to a number of transcription factors. Inhibits the transcriptional activation mediated by PAX5, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with dominant-negative AES.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLE4
Applications Western Blot
Classification Monoclonal
Clone 384CT24.1.3.4
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TLE4
Gene Accession No. Q04727
Gene Alias 5730411M05Rik; AA792082; B lymphocyte gene 1; BCE1; BCE-1; E(spI); enhancer of split groucho 4; ESG; ESG4; Esp2; ESTM13; ESTM14; GRG4; Grg-4; groucho-related protein 4; KIAA1261; Protein ESP2; TLE family member 4, transcriptional corepressor; Tle4; transducin like enhancer of split 4; transducin-like enhancer of split 4; transducin-like enhancer of split 4 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 4, E(spl) homolog; transducin-like enhancer of split 4, homolog of Drosophila E(sp1); transducin-like enhancer of split 4, homolog of Drosophila E(spl); transducin-like enhancer protein 4; X83332; X83333
Gene Symbols TLE4
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 262-290 amino acids from the Central region of human TLE4.
Purification Method Euglobulin Precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.