Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982210
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2210 Supplier Bioss Supplier No. BS12403R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcriptional corepressor that binds to a number of transcription factors. Inhibits the transcriptional activation mediated by PAX5, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with dominant-negative AES.

Specifications

Antigen TLE4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TLE4
Gene Accession No. Q04727
Gene Alias 5730411M05Rik; AA792082; B lymphocyte gene 1; BCE1; Bce-1; BOS_8897; E(spI); enhancer of split groucho 4; ESG; ESG4; Esp2; ESTM13; ESTM14; GRG4; Grg-4; Groucho-related protein 4; KIAA1261; Protein ESP2; TLE4; transducin like enhancer of split 4; transducin-like enhancer of split 4; transducin-like enhancer of split 4 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 4, E(spl) homolog; transducin-like enhancer of split 4, homolog of Drosophila E(sp1); transducin-like enhancer of split 4, homolog of Drosophila E(spl); Transducin-like enhancer protein 4; X83332; X83333
Gene Symbols TLE4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TLE4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7091
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.