Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLE6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109241 Supplier Invitrogen™ Supplier No. PA5109241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Mouse ovary tissue lysate, HepG2, A431, A549, SK-Br-3, mouse small intestine tissue, Daudi.

As a member of the subcortical maternal complex (SCMC), plays an essential role for zygotes to progress beyond the first embryonic cell divisions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLE6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TLE6
Gene Accession No. Q9H808, Q9WVB3
Gene Alias 1810057E06Rik; Grg6; Groucho-related protein 6; PREMBL; TLE6; transducin like enhancer of split 6; transducin-like enhancer of split 6 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 6, homolog of Drosophila E(spl); transducin-like enhancer protein 6
Gene Symbols TLE6
Host Species Rabbit
Immunogen Synthetic peptide within mouse Tle6 aa 157-206.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114606, 79816
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.