Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLK1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298826R
Description
TLK1 Polyclonal specifically detects TLK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
TLK1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 550 | |
EC 2.7.11, EC 2.7.11.1, KIAA0137serine/threonine-protein kinase tousled-like 1, PKU-beta, SNARE protein kinase SNAK, tousled-like kinase 1serine threonine protein kinase | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human TLK1. | |
0.1 mL | |
Apoptosis, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Repair, Tumor Suppressors | |
9874 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction