Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLK1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177982 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177982 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177982 Supplier Novus Biologicals Supplier No. NBP177982
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TLK1 Polyclonal specifically detects TLK1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TLK1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.6 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, ELISA 1:10000-1:50000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. NP_036422
Gene Alias EC 2.7.11, EC 2.7.11.1, KIAA0137serine/threonine-protein kinase tousled-like 1, PKU-beta, SNARE protein kinase SNAK, tousled-like kinase 1serine threonine protein kinase
Gene Symbols TLK1
Host Species Rabbit
Immunogen This affinity purified TLK1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a C-Terminal region near amino acids 730-755 of human TLK1 (Tousled-like kinase) (isoform 1) protein. (Uniprot: Q9UKI8)
Molecular Weight of Antigen 86.7 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9874
Test Specificity This affinity purified antibody is directed against human TLK1 protein. This antibody reacts with both phosphorylated and non-phosphorylated forms of TLK1 at the S743 residue (isoform 1) of the protein. The antibody will react with all isoforms of TLK1. A BLAST analysis was used to suggest reactivity with this protein from human, chimpanzee, orangutan, and dog sources based on 100% homology for the immunogen sequence. Cross reactivity with TLK1 protein from mouse, rat and chicken may occur as this sequence only varies by one amino acid residue (93% homology). Cross reactivity with TLK1 homologues from other sources has not been determined.
Target Species Human, Canine, Chimpanzee
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.