Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLK2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299818JF549
Description
TLK2 Polyclonal antibody specifically detects TLK2 in Human samples. It is validated for ELISASpecifications
TLK2 | |
Polyclonal | |
ELISA | |
EC 2.7.11, EC 2.7.11.1, MGC44450, PKU-ALPHA, serine/threonine-protein kinase tousled-like 2, tousled-like kinase 2serine/threonine kinase | |
Produced in rabbits immunized with purified, recombinant Human TLK2 (Uniprot#: Q86UE8-1; Leu397-Asn772) | |
0.1 mL | |
Cell Cycle and Replication | |
11011 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction