Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLL1 Monoclonal Antibody (4H8C1)
GREENER_CHOICE

Catalog No. PIMA531934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531934 Supplier Invitrogen™ Supplier No. MA531934
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31934 has been tested in indirect ELISA.

This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. A similar protein in mice is required during heart development and specifically processes procollagen C-propeptides and chordin at similar cleavage sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLL1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4H8C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TLL1
Gene Accession No. O43897
Gene Alias ASD6; TLL; TLL1; tolloid like 1; tolloid-like 1; tolloid-like protein 1
Gene Symbols TLL1
Host Species Mouse
Immunogen Purified recombinant fragment of human TLL1 (AA: 870-1013) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.