Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR10 Antibody (28M7A3), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227273AF750
Description
TLR10 Monoclonal specifically detects TLR10 in Human samples. It is validated for Flow Cytometry.Specifications
TLR10 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
TLR10 | |
A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody. | |
Primary | |
Human | |
IgG2a κ |
Flow Cytometry | |
28M7A3 | |
Flow Cytometry | |
CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10 | |
Mouse | |
0.1 mL | |
81793 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction