Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR10 Antibody (28M7A3), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227273G
Description
TLR10 Monoclonal specifically detects TLR10 in Human samples. It is validated for Flow Cytometry.Specifications
TLR10 | |
Monoclonal | |
DyLight 488 | |
Q9BXR5 | |
TLR10 | |
A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody. | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
81793 | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry | |
28M7A3 | |
Flow Cytometry 1ul/1 million cells | |
CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction