Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR10 Antibody (28M7A3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227273MFV450
Description
TLR10 Monoclonal antibody specifically detects TLR10 in Human samples. It is validated for Flow CytometrySpecifications
| TLR10 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 28M7A3 | |
| Flow Cytometry | |
| CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10 | |
| A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 81793 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction