Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR10 Antibody (28M7A3), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227273MFV610
Description
TLR10 Monoclonal antibody specifically detects TLR10 in Human samples. It is validated for Flow CytometrySpecifications
TLR10 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
28M7A3 | |
Flow Cytometry | |
CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10 | |
A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody. | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
81793 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction