Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR10 Antibody (3C10C5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP170343AF350
Description
TLR10 Monoclonal specifically detects TLR10 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
| TLR10 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TLR10 | |
| A partial recombinant protein containing amino acids 20-474 (extracellular domain) of human TLR10. | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 3C10C5 | |
| Flow Cytometry, Flow (Cell Surface) | |
| CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10 | |
| Mouse | |
| 0.1 mL | |
| 81793 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction