Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR10 Antibody (3C10C5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311234
Description
TLR10 Monoclonal specifically detects TLR10 in Human samples. It is validated for Flow Cytometry, Immunoassay.Specifications
TLR10 | |
Monoclonal | |
0.1 mg/ml | |
Flow Cytometry, Immunoassay | |
CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10 | |
The extracellular domain of TLR10 | |
50 μg | |
Cytokine Research, Immunology, Innate Immunity | |
81793 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Flow Cytometry, Immunoassay | |
3C10C5 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction