Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR11 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00

Specifications

Antigen TLR11
Concentration 1.0 mg/mL
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 0.5-1 ug/10-6 cells
Applications Western Blot, Flow Cytometry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056742T
SDP
View Documents
Novus Biologicals
NB10056742SS
0.025 mg
Each for $206.00
Only null left
Add to Cart
 
NB10056742
SDP
View Documents
Novus Biologicals
NB10056742
0.1 mg N/A N/A N/A
Description

Description

TLR11 Polyclonal specifically detects TLR11 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

TLR11
Western Blot 1-3 ug/ml, Flow Cytometry 0.5-1 ug/10-6 cells
Polyclonal
Purified
RUO
Mouse, Rat
NP_991388
442887
A synthetic peptide within amino acid residues 911-926 (CEQLKRRLSKAGQERD) of mouse TLR11 (Genbank accession no. NP_991388) was used as immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Flow Cytometry
Unconjugated
Rabbit
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
PBS with 0.05% Sodium Azide
hgnc31753, Toll like receptor 11
TLR11
IgG
Protein G purified
Immunizing peptide will block the mouse TLR11 band in western blotting.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.