Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR11 Antibody (786404), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7640V
Description
TLR11 Monoclonal antibody specifically detects TLR11 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
TLR11 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
786404 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
hgnc31753, Toll like receptor 11 | |
E. coli-derived recombinant mouse TLR11, Thr209-Gln325, Accession # Q6R5P0 | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
442887 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction