Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLR11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520057
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520057 Supplier Invitrogen™ Supplier No. PA520057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is RAW264.7 cell lysate. PA5-20057 can be used with blocking peptide PEP-0176.

The Toll-like receptor (TLR) family in mammal comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Like its counterparts in Drosophila, TLRs signal through adaptor molecules (1). The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition (2). TLR11 does not respond to known TLR ligands but instead respond specifically to uropathogenic bacteria. TLR11-deficient mice are highly susceptible to infection of the kidneys by uropathogenic bacteria, indicating that TLR11 may play a protective role in preventing infection of internal organs of the urogenital system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Tlr11
Gene Accession No. Q6R5P0
Gene Alias Gm1365; Gm287; RGD1564460; RP23-392K10.4; Tlr11; Tlr12; Toll-like receptor 11; Toll-like receptor 12
Gene Symbols Tlr11
Host Species Rabbit
Immunogen A peptide corresponding to 16 amino acids between the middle and carboxy terminus of mouse TLR11.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239081
Target Species Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.