Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR12 Antibody (1229C) [Janelia Fluor™ 646], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB8086J
Description
TLR12 Monoclonal antibody specifically detects TLR12 in Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
TLR12 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
100131451 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry | |
1229C | |
Western Blot, Flow Cytometry | |
toll-like receptor 12 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TLR12, Thr22-Lys709, Accession # Q6QNU9 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction