Learn More
Invitrogen™ TLR12 Monoclonal Antibody (15F1215)

Description
Based on sequence homology, this antibody may also detect mouse TLR11.
Specifications
Specifications
| Antigen | TLR12 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 15F1215 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites, PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | Tlr12 |
| Gene Accession No. | Q6QNU9 |
| Gene Alias | Gm1365; Gm287; MGC129352; MGC129353; RGD1559145; Tlr11; Tlr12; Toll-like receptor 11; Toll-like receptor 12 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.