Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLR13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523107
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523107 Supplier Invitrogen™ Supplier No. PA523107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The amino acid sequence used as immunogen for this p65 NLS antibody is widely conserved in many species and isoforms including human, mouse, rat, rhesus monkey, chimp, bovine and horse.

TLR13, also known as Toll like receptor 13, is a member of the Toll-like receptor (TLR) family of transmembrane proteins comprised of multiple copies of leucine rich repeats in the extracellular domain and a TIR domain. The exact function of the 991 amino acidTLR13 protein is yet unknown, but reports suggest that it is closely related to TLR3 and participates in the innate immune response to microbial agents. TLR13 binds to and acts via MYD88 and TRAF6, leading to NF-kappa-B activation, cytokine secretion and inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR13
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Tlr13
Gene Accession No. Q6R5N8
Gene Alias AI666735; Gm713; RGD1562655; RP23-186F4.4; Tlr13; Toll-like receptor 13
Gene Symbols Tlr13
Host Species Rabbit
Immunogen A portion of amino acid 800 to 850 of mouse TLR13.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279572, 317227
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.