Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR2 Antibody (67N17F8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280980
Description
TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TLR2 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot : 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin : 1:10-1:500 | |
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
67N17F8 | |
Unconjugated | |
PBS | |
TLR2 | |
Within the range of amino acids 227-398 of human TLR2 were used as the immunogen for the antibody. | |
0.1 mg | |
Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
7097 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction