Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR2 Antibody (67N17F8), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225244FR
Description
TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TLR2 | |
Monoclonal | |
DyLight 680 | |
NP_003255.2 | |
TLR2 | |
Within the range of amino acids 227-398 of human TLR2 were used as the immunogen for the antibody. | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7097 | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
67N17F8 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This product reacts with Human. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction