Learn More
Description
Specifications
Specifications
| Antigen | TLR2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry reported in scientific literature (PMID 31285077), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30618413), Immunoprecipitation reported in scientific literature (PMID 31383741), Immunohistochemistry-Paraffin 5 ug/ml |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | O60603 |
| Gene Alias | CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 |
| Gene Symbols | TLR2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
