Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$215.00 - $454.50

Specifications

Antigen TLR2
Concentration 1.0 mg/mL
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 28819109)
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056573T
SDP
View Documents
Novus Biologicals
NB10056573SS
0.025 mg
Each for $215.00
Only null left
Add to Cart
 
NB10056573
SDP
View Documents
Novus Biologicals
NB10056573
0.1 mg
Each for $454.50
Only null left
Add to Cart
 
Description

Description

TLR2 Polyclonal specifically detects TLR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TLR2
Western Blot 0.5-2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 28819109)
Polyclonal
Purified
RUO
Human, Mouse
NP_003255
7097
This antibody was developed against an extracellular domain of mouse TLR2 (amino acids 564-580 and 751-770).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Immunology, Innate Immunity, Signal Transduction
PBS with 0.05% Sodium Azide
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
TLR2
IgG
Protein G purified
An 86 kDa band is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.