Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR2 Antibody (TL2.1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 61 publication
$207.00 - $524.00
Specifications
Antigen | TLR2 |
---|---|
Clone | TL2.1 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 1 ug/10^6 cells, ELISA reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 ug, Functional reported in scientific literature (PMID 25412776), In vitro assay reported in scientific literature (PMID 24512642), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (PMID 24836676), Block/Neutralize reported in scientific literature (PMID 25412776) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Functional Assay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056722T
![]() |
Novus Biologicals
NB10056722SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB10056722
![]() |
Novus Biologicals
NB10056722 |
0.1 mg |
Each for $524.00
|
|
|||||
Description
TLR2 Monoclonal specifically detects TLR2 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional, In vitro assay, Dot Blot, Flow (Cell Surface), Block/Neutralize.Specifications
TLR2 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Functional Assay | |
Unconjugated | |
Mouse | |
Immunology, Innate Immunity, Signal Transduction | |
O60603 | |
7097 | |
This antibody was raised by immunizing mice with CHO cells transfected with human TLR2 cDNA (Flo et al, 2000). The hybridoma supernatants were selected by flow cytometry. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
TL2.1 | |
Western Blot 1 - 2 ug/ml, Flow Cytometry 1 ug/10^6 cells, ELISA reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 ug, Functional reported in scientific literature (PMID 25412776), In vitro assay reported in scientific literature (PMID 24512642), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (PMID 24836676), Block/Neutralize reported in scientific literature (PMID 25412776) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Canine, Rabbit | |
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
TLR2 | |
IgG2a | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title