Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR2 Antibody (TL2.1), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312062H
Description
TLR2 Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Western BlotSpecifications
TLR2 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
TL2.1 | |
Western Blot | |
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
This antibody was raised by immunising BALB/c mice with CHO cells transfected with human TLR2 cDNA. | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
7097 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction