Learn More
Description
Specifications
Specifications
| Cell Line | Human TLR2/CD282 transfected cell lysate |
| Quantity | 220 μL |
| Storage Buffer | 1X SDS sample buffer: 60mM Tris-Hcl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol |
| Host Species | Human |
| Content And Storage | -80°C in the dark. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| For Use With (Application) | Western Blot |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
