Learn More
Invitrogen™ TLR2 Monoclonal Antibody (mT2.4)

Description
MA1-40080 detects TLR2 from mouse samples. MA1-40080 inhibits murine TLR2-mediated cell activation. It masks the lipopeptide binding site in the recombinant extracellular domain of TLR2 (TLR2ECD) and does not hinder the receptor complex formation. MA1-40080 reacts with the native TLR2 protein and not with denatured TLR2 protein . Furthermore the antibody stains overexpressed, as well as endogenous cell surface- and intracellular mouse TLR2. MA1-40080 has been successfully used in flow cytometry, ELISA, function assays and immunohistochemistry (paraffin and frozen) applications. The MA1-40080 immunogen is mouse TLR2. Endotoxin <24 EU/mg.
Specifications
Specifications
| Antigen | TLR2 |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | mT2.4 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene | Tlr2 |
| Gene Accession No. | Q9QUN7 |
| Gene Alias | CD282; Ly105; TIL4; Tlr2; TLR-2; toll like receptor 2; toll/interleukin-1 receptor-like protein 4; Toll-like receptor; toll-like receptor 2; toll-like receptor 2 variant 1; toll-like receptor 2 variant 2; toll-like-receptor 2, exon 1a_; Toll-like-receptor, exon 1b; Toll-like-receptor, exon 1b_; Toll-like-receptor2, exon 1a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.