Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Mouse anti-Human, Clone: 1030A5.138 , Abnova™

Catalog No. 89935014 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-014 Supplier Abnova Corporation Supplier No. MAB12658

Mouse monoclonal antibody raised against human TLR2.

The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is expressed most abundantly in peripheral blood leukocytes, and mediates host response to Gram-positive bacteria and yeast via stimulation of NF-kappaB. [provided by RefSeq]

Specifications

Antigen TLR2
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1030A5.138
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human TLR2.
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide).
Gene Alias CD282/TIL4
Gene Symbols TLR2
Host Species Mouse
Immunogen A synthetic peptide corresponding to extracellular region of human TLR2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7097
Test Specificity In Ramos, an 86 kD band is observed.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.