Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR3 Antibody (002), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290455DL594
Description
TLR3 Monoclonal specifically detects TLR3 in Mouse samples. It is validated for ELISA.Specifications
TLR3 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
2 | |
ELISA | |
CD283, CD283 antigen, toll-like receptor 3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TLR3 (NP_569054.2; Met1-Leu705). | |
0.1 mL | |
Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Necroptosis, Signal Transduction | |
7098 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction