Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR3 Antibody (002), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290455MFV450
Description
TLR3 Monoclonal antibody specifically detects TLR3 in Mouse samples. It is validated for ELISASpecifications
TLR3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
002 | |
ELISA | |
CD283, CD283 antigen, toll-like receptor 3 | |
Rabbit immunized with purified, recombinant Mouse TLR3 (Accession#: NP_569054.2, Met1-Leu705) | |
0.1 mL | |
Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
7098 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction