Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR3 Antibody (27N3D4), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP227405AF405
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
TLR3 | |
Monoclonal | |
Alexa Fluor 405 | |
CD283, CD283 antigen, toll-like receptor 3 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry | |
27N3D4 | |
Western Blot, Flow Cytometry | |
TLR3 | |
A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody. | |
0.1 mL | |
Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7098 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction