Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR3 Antibody (27N3D4), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP227405MFV610
Description
TLR3 Monoclonal antibody specifically detects TLR3 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
TLR3 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
27N3D4 | |
Flow Cytometry | |
CD283, CD283 antigen, toll-like receptor 3 | |
A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody. | |
0.1 mL | |
Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
7098 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction