Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR3 Antibody (40C1285.6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 75 publications
$236.98 - $532.32
Specifications
| Antigen | TLR3 |
|---|---|
| Clone | 40C1285.6 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 2-4 ug/ 1x10^6 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2 ug/ 1x10^6 cells, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) reported in scientific literature (PMID 24836676), Bioactivity reported in scientific literature (PMID 28894449), Block/Neutralize reported in scientific literature (PMID 28894449) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Neutralization |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224875SS
![]() |
Novus Biologicals
NBP224875SS |
0.025 mg |
Each for $236.98
|
|
|||||
NBP224875
![]() |
Novus Biologicals
NBP224875 |
0.1 mg |
Each for $532.32
|
|
|||||
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Bioactivity, Block/Neutralize.Specifications
| TLR3 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Neutralization | |
| Unconjugated | |
| Mouse | |
| Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors | |
| O15455 | |
| 7098 | |
| A synthetic peptide corresponding to amino acids 55-85 of human TLR3 was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 40C1285.6 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 2-4 ug/ 1x10^6 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2 ug/ 1x10^6 cells, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) reported in scientific literature (PMID 24836676), Bioactivity reported in scientific literature (PMID 28894449), Block/Neutralize reported in scientific literature (PMID 28894449) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Canine | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| TLR3 | |
| IgG1 κ | |
| Protein G purified | |
| This antibody is reactive to Dog, Human and Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title