Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR3 Antibody (619F7.06), Alexa Fluor™ 546, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0470A546100
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TLR3 | |
Monoclonal | |
0.5mg/mL | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
CD283, CD283 antigen, toll-like receptor 3 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine | |
IgM |
Immunocytochemistry, Immunofluorescence | |
619F7.06 | |
Alexa Fluor 546 | |
NP_003256 | |
TLR3 | |
TLR3/CD283 was used as immunogen. | |
0.1 mg | |
Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7098 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction