Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | TLR3 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224564SS
![]() |
Novus Biologicals
NBP224564SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224564
![]() |
Novus Biologicals
NBP224564 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
TLR3 Polyclonal specifically detects TLR3 in Human, Mouse samples. It is validated for Western Blot.Specifications
TLR3 | |
Western Blot 1-3 ug/ml | |
Polyclonal | |
Rabbit | |
Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
7098 | |
A portion of amino acids 750-800 of human TLR3 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse | |
CD283, CD283 antigen, toll-like receptor 3 | |
TLR3 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, chimpanzee, orangutan, macaque and zebrafish, 94% homologous in cow, 90% homologous in opossum and 88% homologous in chicken, horse, platypus, rat, mouse and dog. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title