Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Antibody (76B357.1) - Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 25 publications
$246.61 - $579.29
Specifications
| Antigen | TLR4 |
|---|---|
| Clone | 76B357.1 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5ug/ml, Immunohistochemistry-Frozen 1:500, Flow (Cell Surface) reported in multiple pieces of scientific literature, Flow (Intracellular) reported in scientific literature (PMID 18577706), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready, Knockout Validated, KnockDown Validated, Block/Neutralize reported in scientific literature (PMID 26063617) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Neutralization |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227149SS
![]() |
Novus Biologicals
NBP227149SS |
0.025 mg |
Each for $246.61
|
|
|||||
NBP227149
![]() |
Novus Biologicals
NBP227149 |
0.1 mg |
Each for $579.29
|
|
|||||
Description
TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Rat, Porcine, Bovine, Mammal samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready, Knockout Validated, Knockdown Validated, Block/Neutralize.Specifications
| TLR4 | |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Neutralization | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| O00206 | |
| 7099 | |
| This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 76B357.1 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5ug/ml, Immunohistochemistry-Frozen 1:500, Flow (Cell Surface) reported in multiple pieces of scientific literature, Flow (Intracellular) reported in scientific literature (PMID 18577706), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready, Knockout Validated, KnockDown Validated, Block/Neutralize reported in scientific literature (PMID 26063617) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Mammalia | |
| ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
| TLR4 | |
| IgG2b κ | |
| Protein G purified | |
| The amino acid sequence used has 100% identity in human, chimp, baboon, Chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title