Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Antibody (76B357.1), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056566PEATT594
Description
TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Rat, Porcine, Bovine, Mammal samples. It is validated for Flow Cytometry, Knockout Validated, Knockdown Validated.Specifications
TLR4 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
TLR4 | |
This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564). | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Mammalia | |
IgG2b κ |
Flow Cytometry, Immunoassay, KnockDown | |
76B357.1 | |
Flow Cytometry, Knockout Validated, KnockDown Validated | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7099.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction