Learn More
Description
Specifications
Specifications
| Antigen | TLR4 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Functional Assay, In vitro Assay, CyTOF, Neutralization |
| Classification | Monoclonal |
| Clone | HTA125 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 2 ug/ml. Use reported in scientific literature (PMID 28546218), Flow Cytometry 0.25 ug/ 10^6 cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27744428), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24529879), Functional reported in scientific literature (PMID 25412776), In vitro assay reported in scientific literature (PMID 24676500), Flow (Cell Surface) reported in scientific literature (PMID 17015691), Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 26610398), CyTOF-ready, Block/Neutralize |
| Gene Accession No. | O00206 |
| Gene Alias | ARMD10, CD284, CD284 antigen, hta125, hta125 tlr4, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 |
| Gene Symbols | TLR4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
