Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Antibody (HTA125), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056727V
Description
TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot (Negative).Specifications
TLR4 | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry 1 μL/1 million cells, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Western Blot (Negative) | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
The has been shown to block the activation of monocytes with LPS (Paik, et al, 2003). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot, In vitro Assay, Blocking Assay | |
HTA125 | |
DyLight 405 | |
O00206 | |
TLR4 | |
This antibody was developed by immunizing mice with Ba/F3 cell line expressing human TLR4 cell surface antigen. | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
7099 | |
Human, Mouse, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction