Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Antibody (MTS510), Janelia Fluor™ 549, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP224865JF549
Description
TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry.Specifications
TLR4 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TLR4 | |
This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000). | |
0.1 mL | |
Primary | |
This antibody preferentially recognizes TLR4-MD-2 complex than of TLR4 alone. | |
Store at 4°C in the dark. |
Flow Cytometry | |
MTS510 | |
Flow Cytometry | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Rat | |
Protein G purified | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7099 | |
Mouse | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction