Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Antibody (MTS510), Novus Biologicals™

Rat Monoclonal Antibody has been used in 12 publications
Supplier: Novus Biologicals NB10056560
Description
TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Flow (Cell Surface).Specifications
| TLR4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1-2 ug/10^6 cells, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22573811), Immunoprecipitation 1:10-1:500, Flow (Cell Surface) 1 - 2 ug/ 10^6 cells | |
| ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, mts510, mts510 tlr4, TOLL, toll-like receptor 4 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody preferentially recognizes TLR4-MD-2 complex than of TLR4 alone. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| MTS510 | |
| Unconjugated | |
| O00206 | |
| TLR4 | |
| This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000). | |
| 0.1 mg | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7099 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction