Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$206.50 - $499.50
Specifications
| Antigen | TLR4 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224821SS
![]() |
Novus Biologicals
NBP224821SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224821
![]() |
Novus Biologicals
NBP224821 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
TLR4 Polyclonal specifically detects TLR4 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| TLR4 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| O00206 | |
| 7099 | |
| This antibody was developed against a sythetic peptide corresponding to amino acids 420-456 of human TLR4. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
| TLR4 | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title