Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 22 publications
Supplier: Novus Biologicals NB10056580SS
Description
TLR4 Polyclonal specifically detects TLR4 in Human, Mouse, Rat, Rhesus Macaque samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Proximity Ligation Assay.Specifications
TLR4 | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
This antibody was tested against partial recombinant mouse TLR4 (extracellular portion), and a 75-80 kDa band was observed. Full-length TLR4 is observed at approximately 90 kD. 2. Zager et al (2006, Fig. 10) observed TLR4 as an ∼95 kDa doublet which is thought to represent different degrees of glycosylation. See Product Citation (Renal Physiol 291:F546-F556) for additional information. 3. Presumptive TLR4 30 and 60 kDa cleavage fragments are described in Zagar et al 2006. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In situ PLA | |
1.0 mg/ml | |
Western Blot 2 - 5 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 2 - 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Proximity Ligation Assay | |
O00206 | |
TLR4 | |
This antibody was developed against a sythetic peptide corresponding to amino acids in a range between 30-80 of mouse TLR4. | |
0.025 mg | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
7099 | |
Human, Mouse, Rat, Rhesus Macaque | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction