Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLR4/MD-2 Complex Monoclonal Antibody (MTS510)
GREENER_CHOICE

Catalog No. PIMA516210
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516210 Supplier Invitrogen™ Supplier No. MA516210
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Suggested positive control: Raw or thioglycolate-elicited mouse macrophage cells.

MD-2 is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs are signaling molecules that recognize different pathogen-associated molecular patterns (PAMPs) and serve as an important link between the innate and adaptive immune responses. TLR4, the major signaling receptor for lipopolysaccharide (LPS), requires the binding of MD-2 to its extracellular region for maximal response to LPS. The specificity of this response is determined by the species of MD-2; e.g., human MD-2 transfected into mouse cells can cause mouse TLR4 to react to LPS analogs that are normally antagonistic to human but not mouse TLR4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR4/MD-2 Complex
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone MTS510
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene Tlr4
Gene Accession No. O00206, Q9JHF9, Q9QUK6, Q9Y6Y9
Gene Alias ARMD10; CD284; Esop1; ESOP-1; hToll; lipopolysaccharide response; Lps; Ly87; Ly96; Ly-96; Lymphocyte antigen 96; Md2; MD-2; myeloid differentiation factor-2; myeloid differentiation protein-2; protein MD-2; Ran/M1; Rasl2-8; Tlr4; TLR-4; TOLL; Toll-like receptor 4
Gene Symbols Ly96, Tlr4
Host Species Rat
Immunogen Ba/F3 cell line expressing mouse TLR4 and MD-2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17087, 21898, 23643, 7099
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.