Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Mouse anti-Human, Mouse, Clone: 3G9A4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TLR4 |
---|---|
Clone | 3G9A4 |
Concentration | 0.77 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6781
![]() ![]() |
Proteintech Group Inc
663501IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TLR4 is a member of the Toll like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. It has been implicated in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria.Specifications
TLR4 | |
0.77 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARMD10, CD284, hToll, TLR4, TOLL, toll like receptor 4 | |
TLR4 | |
IgG2b | |
Protein A | |
Antibody |
3G9A4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O00206, Q9QUK6 | |
21898, 7099 | |
TLR4 Fusion Protein Ag13857 | |
Primary | |
-20°C | |
TLR4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title