Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Antibody (10D8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | TLR5 |
---|---|
Clone | 10D8 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
TLR5 Monoclonal antibody specifically detects TLR5 in Human samples. It is validated for Western Blot, ELISASpecifications
TLR5 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
A synthesized peptide derived from Human TLR5 [UniProt O60602] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10D8 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cytokine Research, Immunology, Innate Immunity, Prostate Cancer | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7100 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title