Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Antibody (19D759.2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224787MFV610
Description
TLR5 Monoclonal antibody specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Flow Cytometry, Flow (Cell Surface), Flow (Intracellular)Specifications
TLR5 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Canine | |
Purified |
Flow Cytometry | |
19D759.2 | |
Flow Cytometry, Flow (Cell Surface), Flow (Intracellular) | |
MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5 between amino acids 700-800. The antibody also reacts with mouse TLR5. | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Prostate Cancer | |
7100 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction