Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR5 Antibody (624915) [FITC], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6704F
Description
TLR5 Monoclonal antibody specifically detects TLR5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
TLR5 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
624915 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
E. coli-derived recombinant human TLR5, Ile21-Phe115, Accession # O60602 | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Prostate Cancer | |
7100 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction